Please login to your online account to display your discounted pricing
Wheaton™ Volumetric Modules for Acura™ Electro 926 Regular Shaft Length Pipets
Permanently store performance data in instrument's memory
Manufacturer: wheaton™  WW870114
 View more versions of this product
Catalog No. 03-410-180
  • / Case
Description & Specifications
Specifications
Number of Channels 1
Volume (Metric) Range 0.5 to 10μL
For Use With (Equipment) Acura™ Electro™ 926XS regular shaft length single channel pipette
Tip Style Ultra 10μL
Volume (Metric) 0.5 to 10μL
Length (English) Regular Length
Increments 0.05μL
Accuracy ±1.2%
Compatible Tips Ultra 10μL pipette tip
Finger Style Ergonomic