Please login to your online account to display your discounted pricing

Wheaton™ Vial File™ with Screw Cap: Amber

Amber sample vials with a variety of screw cap options

$163.09 - $248.03

Specifications

Certifications/Compliance USP Type I
Color Amber
Material Borosilicate Glass
Type Sample Vial
For Use With (Application) Used for storing and transporting vials
View More Specs
Products
Catalog Number Mfr. No. Closure Type Diameter (Metric) Outside Volume (English) Volume (Metric) Price Quantity    

02-912-591

 
wheaton™
WW224954
Open top screw cap 12mm 0.405 dr. 1.5mL Case of 60 Each for $248.03

02-912-596

 
wheaton™
WW224897
Screw cap 15mm 1.08 dr. 4mL Case of 60 Each for $163.09

02-912-597

 
wheaton™
WW224955
Open top screw cap 15mm 1.08 dr. 4mL Case of 60 Each for $195.03
Description & Specifications

Specifications

Certifications/Compliance USP Type I
Color Amber
Material Borosilicate Glass
Type Sample Vial
For Use With (Application) Used for storing and transporting vials
Material Closure Phenolic

The Vial File provides a compact, easily accessible means of sample storage. The vials are stored in individual partitions within a clear polystyrene case. Foam inserts provide extra protection for the vials.

  • Alpha numerical indexing card allows for identification of each sample vial
  • Vials are manufactured from borosilicate glass which provides superior chemical resistance
  • Cat. Nos. 02-912-591 and -597 feature open-top black phenolic screw cap for easy access with a syringe
  • The Teflon-faced silicone lined screw cap is ideal for low-temperature applications; provides excellent chemical barrier
  • Vials and caps are autoclavable