Please login to your online account to display your discounted pricing

Wheaton™ V Vial™ with Open-Top Screw Cap

Conical interior bottoms are designed to provide total downward drainage and maximum retrieval of contents by syringe

$332.50 - $614.21

Specifications

Color Amber
Material Borosilicate Glass
For Use With (Application) Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Shape Conical V
View More Specs
Includes:  Silicone-lined, black phenolic screw caps faced with PTFE.
Products
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outside Height (Metric) Price Quantity    

03-410-038

 
wheaton™
W986359NG
5mL 20mm 65mm Case of 12 Each for $614.21

03-410-026

 
wheaton industries
W986354NG
1mL 13mm 44mm Case of 12 Each for $545.94

03-084-108

 
wheaton™
W986356NG
2.0mL 13mm 44mm Case of 12 Each for $332.50
Description & Specifications

Specifications

Color Amber
Material Borosilicate Glass
For Use With (Application) Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Shape Conical V

Made of “320” Type I glass for light-sensitive materials.