Please login to your online account to display your discounted pricing
Wheaton™ Unlined Aluminum Open-Top Seals
Open-top seal exposes stopper for sample retrieval with a syringe
Manufacturer: wheaton™  22417807
 View more versions of this product
Catalog No. 11-126-27
  • / Case of 1000 Each
Description & Specifications
Specifications
Thread Size 20mm
Closure Material Aluminum
Color Green
Material Closure Aluminum
Closure Type Open Top
Quantity 1000 Case
Type Seal
  • Unlined
  • Closure Type: Seal
Accessories
Accessories for Catalog Number: 1112627
Item Mfr. No. Item Description Price Per Unit Quantity  
03-375-24A
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
thermo scientific™
C4020100
Achieve secure, repeatable crimps or remove seals with minimized risk of damage. Each for $469.08