Please login to your online account to display your discounted pricing
Wheaton™ Unlined Aluminum Open-Top Seals
Open-top seal exposes stopper for sample retrieval with a syringe
Manufacturer: wheaton™  22417806
 View more versions of this product
Catalog No. 11-126-26
  • / Case of 1000 Each
Description & Specifications
Specifications
Closure Material Aluminum
Quantity 1000 Case
Type Seal
Thread Size 20mm
Closure Type Open Top
Color Red
Material Closure Aluminum
Accessories
Accessories for Catalog Number: 1112626
Item Mfr. No. Item Description Price Per Unit Quantity