DWK Life Sciences Wheaton™ Unlined Aluminum Center Disc Tear-Out Seals
Exposes the top of the rubber stopper to the syringe when the center disc is opened or removed
Manufacturer: DWK Life Sciences 22418206
Description
Exposes the top of the rubber stopper to the syringe when the center disc is opened or removed
- Available in 13 or 20mm O.D.
Specifications
Red | |
Aluminum | |
1000/Cs. |
13 mm | |
Center Disc Tear-out |