Wheaton™ Trypsinizing Flasks with Side Arm
Manufacturer:  Wheaton™ 355753
 View more versions of this product
Catalog No. 10-042-11B
Specifications Description & Specifications
Specifications
Yes
Screw
Used for forming cell suspensions through trypsin digestion of connective proteins
20-415 Sidearm cap and top cap have white Styrene Butadiene Rubber liner
75mL
33mm
Style: Erlenmeyer Trypsinizing
No
430
Borosilicate Glass