Please login to your online account to display your discounted pricing

Wheaton™ Trypsinizing Flasks


Deep fluted sides enhance agitation

Manufacturer: wheaton™  W355444

 View more versions of this product

Catalog No. 10-042-10C

  • / Each
 

Description & Specifications

Specifications

Thread Size 430
Closure Type Screw
Closure Size 38mm
Material Borosilicate Glass
Capacity (Metric) 150mL
For Use With (Application) Used for forming cell suspensions through Trypsin digestion of connective Proteins
Style Erlenmeyer Trypsinizing
Graduated Yes
Autoclavable Yes