missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ TPE Sterile Connections Kits For Pharma and Biotech Use: Bulk Tubing

Specifically formulated for use with media found in pharmaceutical and biotech processing

Manufacturer:  Wheaton™ WW374738

 View more versions of this product

Catalog No. 03-410-307


 

Specifications Description & Specifications

Specifications

45.72m
USP Class VI
Tubing
6.35mm
Wheaton standard 2.3mm pump head and optional 1.6mm pump head
Yes
50 ft.
0.25 in.
Thermoplastic Elastomer
2.3mm
Yes
2.3mm
0.09 in.