Wheaton™ TPE Sterile Connections Kits For Pharma and Biotech Use
Specifically formulated for use with media found in pharmaceutical and biotech processing
$326.42 - $870.39
Specifications
Type Connection Kit
For Use With (Equipment) Wheaton standard 2.3mm pump head and optional 1.6mm pump head
Includes Male and female luer
Material Thermoplastic Elastomer
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Connection Type Diameter (Metric) Inside Price Quantity  
Catalog Number Mfr. No. Connection Type Diameter (Metric) Inside Price Quantity  
03-410-303 Wheaton™
WW374734
Luer Connectors 6.35mm Case of 5 Each for $831.36
03-410-304 Wheaton™
WW374735
Quick Disconnects 6.35mm Case of 5 Each for $870.39
03-410-344 Wheaton™
W2246220205
Quick Disconnects 3.18mm Pack of 100 Each for $426.22
03-410-345 Wheaton™
W2246220207
Luer Connectors 6.35mm Pack of 100 Each for $398.96
03-410-346 Wheaton™
W22462203
Quick Disconnects 6.35mm Pack of 100 Each for $426.22
36-104-0074 Wheaton™
W374730
Luer Connectors 3.18mm Case for $326.42
Specifications
Description
For use with Wheaton Standard 2.3mm pump head and optional 1.6mm pump head.
  • Radiation sterilized kit includes male and female Luer or quick-disconnects affixed to 5 ft. of TPE tubing
  • Five individually packaged assemblies per case for quick implementation on high value samples, cultures or titers
  • USP CLASS VI and animal derived components free (ADCF)
  • Sterilizable by autoclave, radiation, EtO, chemicals, reusable, heat sealable, heat weldable
Order Info
Bulk tubing available separately (Cat. Nos. 03-410-305 through -307.)
Description & Specifications
Specifications
Connection Kit
Wheaton standard 2.3mm pump head and optional 1.6mm pump head
Male and female luer
Thermoplastic Elastomer