missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ TPE Sterile Connections Kits For Pharma and Biotech Use

Specifically formulated for use with media found in pharmaceutical and biotech processing

Manufacturer:  Wheaton™ W2246220207

 View more versions of this product

Catalog No. 03-410-345


 

Specifications Description & Specifications

Specifications

USP Class VI
Thermoplastic Elastomer
0.06 in.
Yes
Male and female luer
Wheaton standard 2.3mm pump head and optional 1.6mm pump head
6.35mm
0.25 in.
1.52m
Connection Kit
1.6mm
5 ft.
Yes
1.6mm
Luer Connectors