Please login to your online account to display your discounted pricing

Wheaton™ Top-Drive 11-Deck R2P 2.0 Roller Apparatus


Advanced Production Scale Roller Apparatus

Manufacturer: wheaton™  WWRTPF8110A

 View more versions of this product

Catalog No. 02-924-906

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Capacity (English) 85 Bottle positions
Controller Type R2P 2.0
For Use With (Equipment) Roller culture system
Quantity 11
Spacing 6 in.
Format 6 in.