missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Tenbroeck Tissue Grinder

With expanded reservoir and pouring lip

Manufacturer:  DWK Life Sciences 357422

 View more versions of this product

Catalog No. 08-414-13A



Description

Description

With expanded reservoir and pouring lip

  • Precision manufactured from Wheaton-33™ low extractable borosilicate glass
  • Can be operated by hand or slow-speed motor
  • Hollow handle permits packing with ice to reduce heat buildup
  • Clearance between pestle and tube is approximately 0.12mm

Specifications

Specifications

0.06 oz.
Borosilicate Glass
5.51 in.
1.9 in.
0.43 in.
0.06 oz., 2mL
2mL
Tenbroeck tissue grinder
140mm
50mm
11mm
SDS
Documents