Wheaton™ Tapered Tissue Grinders With PTFE Pestles
Tapered tissue grinder with a PTFE pestle for soft tissues
Manufacturer:  Wheaton™ W358141
 View more versions of this product
Catalog No. 02-911-535
Specifications Description & Specifications
Specifications
0.16 oz.
Borosilicate Glass, PTFE
8.6 in.
3.6 in.
0.51 in.
0.16 oz., 5mL
5mL
Tapered tissue grinder
220mm
93mm
13mm