missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Provide airtight covers
Manufacturer: DWK Life Sciences 224100172
Description
Provide airtight covers
- Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
- Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
Specifications
1.3 mm | |
Fits 1.3mm I.D., 20mm O.D. mouth of serum bottles and vials | |
Yes | |
20 mm |
Not for lyophilization | |
45 Durometer | |
Red | |
Rubber |