missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Sleeve Stopper

Provide airtight covers

Manufacturer:  DWK Life Sciences 224100060

 View more versions of this product

Catalog No. 06-406-10A



Description

Description

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications

Specifications

1.3 cm
Fits 0.7cm I.D., 1.3cm O.D. mouth of serum bottles and vials
Yes
5 mm
Rubber
Multi-puncture applications
40 Durometer
Red
11 mm
Sleeve Stopper
SDS
Documents
Product Certifications