missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Snap-on Stopper
Provide airtight covers
Supplier: DWK Life Sciences W224100181
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Gray | |
2 cm | |
Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials | |
Yes |
Snap-On Stopper | |
Chlorobutyl Rubber | |
45 Durometer |