DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Provide airtight covers
Manufacturer:  DWK Life Sciences W224100093
 View more versions of this product
Catalog No. 06-447E
Description
Description
Provide airtight covers
  • Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
  • Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
Specifications
Specifications
7 mm
Fits 7mm I.D., 13mm O.D. mouth of serum bottles and vials
Yes
13 mm
2-Leg Lyophilization Stopper; Butyl Rubber
Lyophilization
50 Durometer
Gray
Butyl Rubber