missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Snap-on Stopper
Provide airtight covers
Manufacturer: DWK Life Sciences 224100080
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
1.3 cm | |
40 Durometer | |
Red | |
Rubber |
Fits 0.7cm I.D., 1.3cm O.D. mouth of serum bottles and vials | |
Yes | |
13 mm |