missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Provide airtight covers
Supplier: DWK Life Sciences 224100192
Description
Provide airtight covers
- Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
- Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
Specifications
Gray | |
13 mm | |
13 mm | |
Chlorobutyl Isoprene Blend | |
50 Durometer |
2-Leg Lyophilization Stopper; Chlorobutyl Isoprene blend | |
Lyophilization | |
20 mm | |
Fits 13mm I.D., 20mm O.D. mouth of serum bottles and vials | |
Yes |