missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Sleeve Stopper
Provide airtight covers
Manufacturer: DWK Life Sciences 224100161
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
2cm | |
Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials | |
Yes | |
20mm |
Multi-puncture applications | |
40 Durometer | |
Red | |
Rubber |