DWK Life Sciences Wheaton™ Columbia Jars for Cover Slips
For staining applications, cleaning cover slips, as well as coating them with materials such as poly-lysine or silane. Wheaton™ Columbia Jars for Cover Slips are manufactured from soda lime glass.
Supplier: DWK Life Sciences W9001806
Description
- Longer cover slips can be accommodated if the cap is removed
- Includes a 43-400 white polypropylene screw cap with a PTFE coated polyethylene liner
- Holds up to 4 cover slips, 17-23mm wide and up to 30mm long
Specifications
White | |
Polypropylene | |
Round | |
Staining Jar |
Closure Included | |
Screw | |
Soda Lime Glass |