DWK Life Sciences Wheaton™ Stainless Steel Microtest Manifold
Make dispensing easier and quicker by selecting a Wheaton Stainless Steel manifold dispensing system.
Manufacturer:  DWK Life Sciences 851382
 View more versions of this product
Catalog No. 13-708-3
Description
Description
  • Unique dispensing system enables performance of sample inoculations and cell feedings faster and more efficiently under sterile conditions
  • Ideal for use with 24 and 96 well microplates
Specifications
Specifications
4
Microplates
Angled
Microtest Manifold
Yes
Stainless Steel
Documents