Please login to your online account to display your discounted pricing
Wheaton™ Stainless Steel Microtest Manifold
Make dispensing easier and quicker by selecting a Wheaton Stainless Steel manifold dispensing system.
Manufacturer: wheaton™  W851382
 View more versions of this product
Catalog No. 02-912-801
  • / Case
Description & Specifications
Specifications
For Use With Microplates
Item Description Angled
Positions 4
Product Type Microtest Manifold
Autoclavable Yes
Material Stainless Steel
  • Unique dispensing system enables performance of sample inoculations and cell feedings faster and more efficiently under sterile conditions
  • Ideal for use with 24 and 96 well microplates