Please login to your online account to display your discounted pricing
Wheaton™ Socorex Calibra™ 832 Digital Macropipettes
Combine mechanical precision, comfort and ease of use
$1,173.00 - $1,337.14
Specifications
Accuracy ±0.5%, <±1%, <±1.5%
Autoclavable Yes
Certifications/Compliance CE, IVD 98/79 EEC, GMP, GLP, NCCLS
Model Digital 832
Finger Style Ergonomic
View More Specs
Products
Catalog Number Mfr. No. Compatible Tips Increments Volume (Metric) Price Quantity    
13-707-77A
  Accessories
wheaton™
851340
Macro 2mL tip 0.01mL 0.2 to 2mL Each for $1,173.00
13-707-77C
  Accessories
wheaton™
851345
Macro 10mL tip 0.1mL 1 to 10mL Each for $1,337.14
Description & Specifications
Specifications
Accuracy ±0.5%, <±1%, <±1.5%
Autoclavable Yes
Certifications/Compliance CE, IVD 98/79 EEC, GMP, GLP, NCCLS
Model Digital 832
Finger Style Ergonomic
For Use With (Application) Gel sequencing and biotechnology applications
UV Monitoring Yes
Warranty 3 Years
  • Unique twin cam system
  • Glove friendly volume setting and tip ejection
  • Fully autoclavable without disassembling pipette
  • Pre-calibrated steps
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.