DWK Life Sciences Wheaton™ Socorex Calibra™ 822 Digital Micropipette
For gel sequencing and biotechnology applications
Manufacturer:  DWK Life Sciences W851168
 View more versions of this product
Catalog No. 13-707-5
Description
Description
  • Single-channel, variable volume with twin cams
  • Precalibrated steps to provide true digital volume entry
  • Color-coded to ensure proper tip selection
Specifications
Specifications
±1.5%, <±0.6%, <±0.5%
CE, IVD 98/79 EEC, GMP, GLP, NCCLS
Gel sequencing and biotechnology applications
5μL
3 Years
100 to 1000μL
Yes
Micro 1000μL tip
Micro 1000μL tip
Yes
Digital 822
Documents
Accessories