DWK Life Sciences Wheaton™ Socorex Calibra™ 822 Digital Micropipette
For gel sequencing and biotechnology applications
Manufacturer:  DWK Life Sciences W851166
 View more versions of this product
Catalog No. 13-707-4
Description
Description
  • Single-channel, variable volume with twin cams
  • Precalibrated steps to provide true digital volume entry
  • Color-coded to ensure proper tip selection
Specifications
Specifications
±1.5%, <±0.9%, <±0.6%
CE, IVD 98/79 EEC, GMP, GLP, NCCLS
Gel sequencing and biotechnology applications
1μL
3 Years
20 to 200μL
Yes
Micro 200μL tip
Micro 200μL tip
Yes
Digital 822
Documents
Accessories