DWK Life Sciences Wheaton™ Socorex Acura™ Single-Channel Fixed-Volume Micropipetters
Separate pushbuttons for dispensing and tip ejection
Manufacturer:  DWK Life Sciences W810002
 View more versions of this product
Catalog No. 13-709-01
Description
Description
Reduce dead volume, avoid accidental de-tipping, and result in an accuracy level far exceeding that of ordinary pipetters.
  • Attractive, comfortable, Swiss-precision design
  • JustTip™adjustable tip ejector allows use of most third-party tips
  • Positive stops differentiate dispense movement from overshoot movement
  • Separate dispensing and tip-ejection buttons can be manipulated easily with just the thumb
  • Maximum accuracy is ensured through calibrated plunger stroke and overshoot movement that flushes the tip empty
  • Positive stops differentiate dispense movement from overshoot movement
  • Autoclavable to 121°C (248°F) without assembly, allowing easier decontamination
Order Info
Compatible with Wheaton Precision Micropipette tips and all Fisherbrand universal-fit tips.
Warranty
Three-years, including two years on original calibration
Specifications
Specifications
±1.4%
ISO8655
Wheaton precision micropipette tip, universal fit tip
3 Years
5μL
Yes
Wheaton precision micropipette tip, universal fit tip
JusTip™ adjustable tip ejection
815
Documents
Accessories