DWK Life Sciences Wheaton™ Shorty Vials™ Glass Sample Vials in Lab File™: without Caps
Come in convenient Lab File partitioned trays
Manufacturer:  DWK Life Sciences 224822
 View more versions of this product
Catalog No. 06-412-6
Description
Description
Come in convenient Lab File partitioned trays
Offering a variety of uses ranging from sample storage to freeze-drying.
  • Clear vials made from borosilicate glass
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Shorty Vials feature a low profile for greater stability and less dead volume
  • Caps sold separately
  • Vials and caps are autoclavable
  • Selected items fit M-T Vial File™—polystyrene trays with partitions that provide an easy way to inventory samples or to store empty vials
Clear Vials
  • Made from low extractable borosilicate glass to provide superior chemical resistance
  • Conform to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Specifications
Clear
Borosilicate Glass (Low Extractable)
For uses ranging from sample storage to freeze drying
1.08 dr.
Screw Cap
USP Type I, ASTM E438, Type I, Class A
19 mm
4 mL
Screw
Autoclavable
43 mm
Vial Rack