Wheaton™ Shaker Flasks With Three Baffles
Manufacturer:  Wheaton™ 353262
 View more versions of this product
Catalog No. 10-042-7D
Specifications
Description
Flasks, Shaker, Glass, Wheaton, With three baffles, 500mL, Break-resistant lip, For use with reciprocating or rotary shakers
  • For use with orbital or reciprocating shakers
  • Baffles improve aeration and mixing for microbial and bacterial cultures
  • Break resistant lip for use with stopper, cotton plug or film cover
  • Pinched neck design to reduce splashing on flasks with 4 baffles
Description & Specifications
Specifications
Yes
Style: Shaker
120mm
6.8 in.
Pinched
Borosilicate Glass
Baffled
4.7 in.
Orbital or reciprocating shakers
17.3cm
500mL