missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Serum Vials
Available as clear and amber vials
Manufacturer: DWK Life Sciences 223683
Description
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
- Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
- Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
- Withstand extreme temperature variations
- Necks finished for aluminum crimp seals
- Blowback feature of neck and specially designed bottom radius for lyophilization applications
- Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Specifications
0.27 in. | |
Clear | |
Borosilicate Glass | |
For lyophilization, packaging, sampling and freeze drying applications | |
Autoclavable | |
32 mm |
7 mm | |
15 mm | |
2 mL | |
0.5 dr. | |
Crimp Cap | |
USP Type I, ASTM E438, Type I, Class A |