DWK Life Sciences Wheaton™ Self-Refilling Repetitive Syringes, Microtest Manifolds
Manufacturer:  DWK Life Sciences 851381
 View more versions of this product
Catalog No. 13-708-2
Description
Description
  • Stainless steel with PTFE stoppers, autoclavable
  • Increased efficiency for inoculations, cell feedings, and washing
  • Ideal for use with 24 or 96 well microplates
  • Tightly fitting Luer and Luer Lock syringe nozzles
  • Available in both a straight T style and an angular style with an offset hub
Specifications
Specifications
±6%
Yes
Stainless Steel
Syringes
Straight
Self refilling repetitive syringes, 24 and 96-well plates, Acura™ 865 microdispenser pipette
8
Documents