Wheaton™ Four Place Biostir Heavy-Duty Magnetic Stirrers
Manufacturer:  Wheaton™ W900703A
 View more versions of this product
Catalog No. 02-927-086
Specifications Description & Specifications
Specifications
UL, CSA, CE, WEEE and RoHs
150 to 1200rpm
15.51 in.
120VAC (North America)
4.0