Please login to your online account to display your discounted pricing
Wheaton™ Four Place Biostir Heavy-Duty Magnetic Stirrers
$3,232.94 - $3,232.94
Specifications
Certifications/Compliance UL, CSA, CE, WEEE and RoHs
Speed 150 to 1200rpm
Stirring Positions 4.0
View More Specs
Products
Catalog Number Mfr. No. Electrical Requirements Price Quantity    
02-927-086
wheaton™
W900703A
120VAC (North America) Each for $3,232.94
Description & Specifications
Specifications
Certifications/Compliance UL, CSA, CE, WEEE and RoHs
Speed 150 to 1200rpm
Stirring Positions 4.0
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.