DWK Life Sciences Wheaton™ Flat Bottom Flasks For Soxhlet Extraction Apparatus
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Supplier:  DWK Life Sciences 428126
 View more versions of this product
Catalog No. 02-912-577
Description
Description
  • Flat bottom flask 24/40
Specifications
Specifications
125 mL
For continuous extraction of analytes from a solid into an organic solvent
Flat
Borosilicate Glass
24/40