missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ E-Z Vial™ Convenience Pack - Amber Vials with 11mm Crimp Finish

Manufacturer:  Wheaton™ W2251720204

 View more versions of this product

Catalog No. 02-926-938


 

Specifications Description & Specifications

Specifications

1.8mL
PTFE/Silicone
Blue
Borosilicate Glass
Amber
Aluminum
32mm
1.25 in.
100 Pack
1.8mL
E-Z Vial™ Convenience Pack
11mm
12mm
Crimp