DWK Life Sciences Wheaton™ BioStir™ Magnetic Stirrers
Ideal for general mixing and difficult-to-dissolve solutes. Wheaton™ BioStir™ Magnetic Stirrers are available in single or 4-place units.
Manufacturer:  DWK Life Sciences W900701A
 View more versions of this product
Catalog No. 02-927-072
Description
Description
  • Programmable
  • 5-200 RPM
  • Remote control and alarm capable
  • Waterproof stainless steel housing
  • Operating temperature range of 15°C to 40°C
Specifications
Specifications
North America
Slow
120VAC
Micro-stir
Documents