missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ 12 x 32mm LVI Vial Convenience Pack With Crimp Top Finish and Amber Vial With Patch

Manufacturer:  Wheaton™ W2253280801

 View more versions of this product

Catalog No. 03-410-461


 

Specifications

Description

Vial; Wheaton; LVI; Type I borosilicate glass made; Seal type: black polypropylene ABC; Septa type: red PTFE/silicone with slit; Color: amber; Vol.: 0.3mL; w/writing patch; Supplied with 11mm aluminum seal and/or 9mm screw cap; 32 x 12

  • Borosilicate glass vial with 300μL glass limited volume insert
  • Insert is fused to vial to prevent needle damage
  • Crimp top seal
  • Writing patch on all vials
Description & Specifications

Specifications

Screw cap
12mm
Borosilicate Glass
0.3mL
Polypropylene
Yes
0.1 dr.
0.3mL
Amber
LVI™ Vial
32mm
PTFE/Silicone
Screw