missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ 12 x 32mm LVI Vial Convenience Pack With Crimp Top Finish and Amber Vial With Patch

Manufacturer:  Wheaton™ W2253280204

 View more versions of this product

Catalog No. 03-410-451


 

Specifications Description & Specifications

Specifications

Screw cap
12mm
Borosilicate Glass
0.3mL
Polypropylene
Yes
0.1 dr.
0.3mL
Amber
LVI™ Vial
32mm
PTFE/Silicone
Screw