DWK Life Sciences Wheaton™ Safety-Coated Narrow Mouth Amber Bottles with Aluminum-Foil-Lined Black Phenolic Caps
Manufacturer:  DWK Life Sciences 220936
 View more versions of this product
Catalog No. 06-451-318
Description
Description
  • Plastisol-coated soda-lime glass
  • Coating is inert and highly resistant to strong acids, alkalis, and most solvents
  • Coating reduces risk of personal injury or contamination
  • Black, foil-lined caps included
  • Not autoclavable
Compliance
Helps meet OSHA requirements for safe handling and disposal of toxic or noxious substances.
Specifications
Specifications
USP Type III
Aluminum Foil
33-400 mm
96 mm
210 mm
1000 mL
Plastisol
3.7 in.
Black
Yes
Phenolic
Screw Cap
8.2 in.
32 oz., 1000 mL
32 oz.
Amber
Plastisol-coated Soda Lime Glass