Please login to your online account to display your discounted pricing
Wheaton™ Screw Neck Glass Diagnostic Bottles
Provides superior chemical resistance. Wheaton™ Screw Neck Glass Diagnostic Bottles are manufactured from Wheaton 400 borosilicate glass that conforms to USP Type I requirements.
$1,903.43 - $1,903.43
Specifications
Certifications/Compliance USP Type I
Closure Type I-Loc; / Open-Top Screw
Closure Size 20-400
Color Amber
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Diameter (English) Height (English) Price Quantity    
06-440-35
wheaton™
219371
0.34 oz., 10mL 0.9 in. 2.4 in. Case of 480 Each for $1,903.43
Description & Specifications
Specifications
Certifications/Compliance USP Type I
Closure Type I-Loc; / Open-Top Screw
Closure Size 20-400
Color Amber
Material Glass
  • Autoclavable
  • Amber-colored glass
  • Can by used for lyophilization
  • Alternative to serum bottles and vials
  • Fits with I-Loc™ closure or screw cap with thin flange stopper
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.