Please login to your online account to display your discounted pricing

Wheaton™ Round Translucent LDPE Bottles


Very flexible, high impact strength plastic

Manufacturer: wheaton™  221142

 View more versions of this product

Catalog No. 03-439-70B

  • / Case of 144 Each
 

Description & Specifications

Specifications

Closure Included Yes
Closure Color White
Closure Material Polyethylene
Color Translucent
Diameter (English) 0.98 in.
Height (Metric) 78mm
Capacity (English) 0.5 oz.
Certifications/Compliance ISO 9001
Closure Screw
Diameter (Metric) 25mm
Height (English) 3.07 in.
Material LDPE
Capacity (Metric) 15mL
Closure Liner Foam Polyethylene
Closure Size 15-415
Neck Finish 15-415
Capacity 0.5 oz., 15mL

Narrow mouth is ideal for liquids.

  • Excellent for mild and strong buffers, good chemical resistance
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.