Please login to your online account to display your discounted pricing

Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers


Unique cone liner design offers superior torque retention and excellent sealing characteristics

Manufacturer: wheaton™  239253

 View more versions of this product

Catalog No. 02-984-15

  • / Case of 144 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Redi-pak™ container
Neck Finish 20-400
Material Phenolic
Color Black
Certifications/Compliance ISO 9001
Closure Liner Polyseal
No. per Case 144 Cs.