Please login to your online account to display your discounted pricing

Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers


Unique cone liner design offers superior torque retention and excellent sealing characteristics

Manufacturer: wheaton™  239260

 View more versions of this product

Catalog No. 02-984-31

  • / Case of 144 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner Polyseal
Color Black
Material Phenolic
No. per Case 144 Cs.
For Use With (Equipment) Redi-pak™ container
Neck Finish 33-400

  • Poly-Seal liner is manufactured from low density polyethylene
  • Phenolic
  • Screw cap
  • Color: Black