Please login to your online account to display your discounted pricing

Wheaton™ Red Polypropylene Caps for Narrow-Mouth Containers


For use with Wheaton leak-resistant narrow mouth bottles

Manufacturer: wheaton™  23950603

 View more versions of this product

Catalog No. 09-877-11C

  • / Case of 72 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Color Red
Material Polypropylene
No. per Case 72 Cs.
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Neck Finish 20-410