missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Red Polypropylene Caps for Narrow-Mouth Containers
For use with Wheaton leak-resistant narrow mouth bottles
$58.83 - $81.34
Specifications
Certifications/Compliance | ISO 9001 |
---|---|
Color | Red |
Material | Polypropylene |
For Use With (Equipment) | Redi-pak™ leak resistant narrow mouth bottle |
Catalog Number | Mfr. No. | Neck Finish | No. per Case | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Neck Finish | No. per Case | Price | Quantity | ||||
09-877-11C
|
DWK Life Sciences
23950603 |
20-410 | 72 Cs. |
Case of 72 Each for $61.77
|
|
09-877-10C
|
DWK Life Sciences
23950103 |
13-425 | 72 Cs. |
Case of 72 Each for $58.83
|
|
09-877-12C
|
DWK Life Sciences
23951003 |
24-410 | 72 Cs. |
Case of 72 Each for $81.34
|
|
Description
- Leak-proof design
- Autoclavable
Specifications
ISO 9001 | |
Red | |
Polypropylene | |
Redi-pak™ leak resistant narrow mouth bottle |