Please login to your online account to display your discounted pricing

Wheaton™ Red Polypropylene Caps for Narrow-Mouth Containers


For use with Wheaton leak-resistant narrow mouth bottles

Manufacturer: wheaton™  23951203

 View more versions of this product

Catalog No. 09-877-13C

  • / Case of 48 Each
 

Description & Specifications

Specifications

Neck Finish 28-410
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Certifications/Compliance ISO 9001
Color Red
Material Polypropylene
No. per Case 48 Cs.