Please login to your online account to display your discounted pricing

Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Red


Manufacturer: wheaton™  W242532A

 View more versions of this product

Catalog No. 03-439-90

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Color Red
Material Polypropylene
No. per Case 1000 Cs.
For Use With (Equipment) Dropping bottle
Neck Finish 13-425