Please login to your online account to display your discounted pricing

Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Red


Manufacturer: wheaton™  W242522A

 View more versions of this product

Catalog No. 03-440-01

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Material Polypropylene
Neck Finish 20-410
Certifications/Compliance ISO 9001
Color Red
For Use With Dropping bottle
No. per Case 1000 Cs.