Please login to your online account to display your discounted pricing

Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Purple


Manufacturer: wheaton™  W242537A

 View more versions of this product

Catalog No. 03-439-84

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Neck Finish 13-425
Certifications/Compliance ISO 9001
Material Polypropylene
Color Purple
No. per Case 1000 Cs.
For Use With Plastic dropping bottle